siRNA Details
| Gene Symbol: NLGN4Y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206850.1 | NP_001193779.1 | 6 | 3086 | ||
| NM_014893.4 | NP_055708.3 | 6 | 3416 | |||
| NR_028319.1 | 6 | 3638 | ||||
| NR_046355.1 | 7 | 3403 | ||||
| XM_006724874.2 | XP_006724937.1 | 7 | 3648 | |||
| XM_011531424.2 | XP_011529726.1 | 7 | 3966 | |||
| XM_011531425.2 | XP_011529727.1 | 7 | 3738 | |||
| XM_011531426.2 | XP_011529728.1 | 7 | 3699 | |||
| XM_011531427.1 | XP_011529729.1 | 7 | 3453 | |||
| XM_011531428.1 | XP_011529730.1 | 8 | 3634 | |||
| XM_011531429.2 | XP_011529731.1 | 7 | 4249 | |||
| XM_011531430.2 | XP_011529732.1 | 7 | 3587 | |||
| XM_017030034.1 | XP_016885523.1 | 6 | 3678 | |||
| XM_017030035.1 | XP_016885524.1 | 6 | 3599 | |||
| XM_017030036.1 | XP_016885525.1 | 6 | 3393 | |||
| XM_017030037.1 | XP_016885526.1 | 6 | 3391 | |||
| XM_017030038.1 | XP_016885527.1 | 6 | 4189 | |||
| XM_017030039.1 | XP_016885528.1 | 7 | 2924 | |||
| XM_017030040.1 | XP_016885529.1 | 6 | 2773 | |||
| GenBank mRNA | AB023168.1 | - | 6 | 2942 | ||
| AF376804.1 | - | 6 | 3416 | |||
| BX537428.1 | - | 6 | 3639 | |||

NLGN4Y
Genomic Map