siRNA Details
| Gene Symbol: PLA2G3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015715.4 | NP_056530.2 | 7 | 2413 | ||
| XM_011530204.1 | XP_011528506.1 | 7 | 2635 | |||
| XM_011530205.1 | XP_011528507.1 | 7 | 2624 | |||
| GenBank mRNA | AF220490.1 | - | 7 | 2413 | ||
| BC025316.1 | - | 7 | 2289 | 5458919 | ||

PLA2G3
Genomic Map