siRNA Details
| Gene Symbol: RRAGB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006064.4 | NP_006055.3 | 7 | 1328 | ||
| NM_016656.3 | NP_057740.2 | 8 | 1412 | |||
| XM_005261973.4 | XP_005262030.1 | 8 | 749 | |||
| XM_017029217.1 | XP_016884706.1 | 7 | 658 | |||
| XM_017029218.1 | XP_016884707.1 | 8 | 1368 | |||
| GenBank mRNA | AK289407.1 | - | 7 | 1261 | ||
| AL831926.1 | - | 7 | 1267 | |||
| BC034726.1 | - | 7 | 1259 | 3919252 | ||
| CD676755.1 | - | 1 | 53 | |||
| X90530.1 | - | 8 | 1203 | |||

RRAGB
Genomic Map