siRNA Details
| Gene Symbol: TM2D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032027.2 | NP_114416.1 | 7 | 962 | ||
| NR_135160.1 | 8 | 1092 | ||||
| NR_135161.1 | 6 | 888 | ||||
| NR_135162.1 | 8 | 1089 | ||||
| GenBank mRNA | AF353990.1 | - | 7 | 961 | ||
| BC029486.1 | - | 7 | 680 | 5271098 | ||
| BC048995.1 | - | 7 | 678 | |||

TM2D1
Genomic Map