siRNA Details
| Gene Symbol: AIFM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018060.2 | NP_001018070.1 | 2, 3 | 248 | ||
| NM_144704.2 | NP_653305.1 | 2, 3 | 248 | |||
| NR_027464.1 | 2, 3 | 315 | ||||
| GenBank mRNA | AK055035.1 | - | 2, 3 | 232 | ||
| AK295581.1 | - | 2, 3 | 218 | |||
| AK316042.1 | - | 2, 3 | 211 | |||
| BC032485.1 | - | 2, 3 | 215 | 5221203 | ||
| CR456342.1 | - | 1, 2 | 67 | |||
| DA396585.1 | - | 2, 3 | 270 | |||
| DB172173.1 | - | 2, 3 | 315 | |||

AIFM3
Genomic Map