siRNA Details
| Gene Symbol: RBMS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003792.2 | NP_001003792.1 | 2 | 600 | ||
| NM_001003793.2 | NP_001003793.1 | 2 | 603 | |||
| NM_001177711.1 | NP_001171182.1 | 2 | 603 | |||
| NM_001177712.1 | NP_001171183.1 | 2 | 603 | |||
| NM_014483.3 | NP_055298.2 | 2 | 603 | |||
| XM_005265060.2 | XP_005265117.1 | 2 | 1052 | |||
| XM_005265061.2 | XP_005265118.1 | 2 | 1055 | |||
| XM_005265062.1 | XP_005265119.1 | 2 | 1055 | |||
| XM_017006178.1 | XP_016861667.1 | 2 | 1052 | |||
| XM_017006179.1 | XP_016861668.1 | 2 | 1052 | |||
| XM_017006180.1 | XP_016861669.1 | 2 | 1055 | |||
| XM_017006181.1 | XP_016861670.1 | 2 | 1052 | |||
| XM_017006182.1 | XP_016861671.1 | 3 | 1131 | |||
| GenBank mRNA | AF023259.1 | - | 2 | 380 | ||
| AK292789.1 | - | 2 | 279 | |||
| AY236871.1 | - | 2 | 463 | |||
| AY236872.1 | - | 2 | 460 | |||
| BC117315.1 | - | 2 | 300 | 40125866 | ||
| BC143521.1 | - | 2 | 300 | 9052030 | ||

RBMS3
Genomic Map