siRNA Details
| Gene Symbol: Cdc42ep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026514.2 | NP_080790.1 | 2 | 1561 | ||
| GenBank mRNA | AK014281.1 | - | 2 | 1563 | ||
| AK019447.1 | - | 2 | 1563 | |||
| AK076782.1 | - | 2 | 1360 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdc42ep3| Gene Symbol: Cdc42ep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026514.2 | NP_080790.1 | 2 | 1561 | ||
| GenBank mRNA | AK014281.1 | - | 2 | 1563 | ||
| AK019447.1 | - | 2 | 1563 | |||
| AK076782.1 | - | 2 | 1360 | |||