siRNA Details
Gene Symbol: Gpr137b-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF154337.1 | - | 7 | 1700 | ||
AK170103.1 | - | 7 | 1710 | |||
AK171335.1 | - | 7 | 1710 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gpr137b-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF154337.1 | - | 7 | 1700 | ||
AK170103.1 | - | 7 | 1710 | |||
AK171335.1 | - | 7 | 1710 |