siRNA Details
| Gene Symbol: UTP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014503.2 | NP_055318.2 | 2, 3 | 287 | ||
| GenBank mRNA | AJ006778.1 | - | 2, 3 | 275 | ||
| AK307934.1 | - | 2, 3 | 287 | |||
| BX957203.1 | - | 2, 3 | 306 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UTP20| Gene Symbol: UTP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014503.2 | NP_055318.2 | 2, 3 | 287 | ||
| GenBank mRNA | AJ006778.1 | - | 2, 3 | 275 | ||
| AK307934.1 | - | 2, 3 | 287 | |||
| BX957203.1 | - | 2, 3 | 306 | |||