siRNA Details
| Gene Symbol: MEX3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032246.4 | NP_115622.2 | 2 | 2410 | ||
| GenBank mRNA | AB095929.1 | - | 1 | 3222 | ||
| AK094145.1 | - | 1 | 1643 | |||
| AK094633.1 | - | 1 | 1074 | |||
| AK131424.1 | - | 2 | 2287 | |||
| AL136778.1 | - | 1 | 1064 | |||
| BC036211.1 | - | 2 | 2408 | 5266259 | ||

MEX3B
Genomic Map