siRNA Details
| Gene Symbol: KRTAP5-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005553.3 | NP_005544.4 | 1 | 923 | ||
| GenBank mRNA | AB126078.1 | - | 1 | 719 | ||
| X63755.1 | - | 1 | 923 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP5-9| Gene Symbol: KRTAP5-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005553.3 | NP_005544.4 | 1 | 923 | ||
| GenBank mRNA | AB126078.1 | - | 1 | 719 | ||
| X63755.1 | - | 1 | 923 | |||