siRNA Details
| Gene Symbol: PDAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014891.6 | NP_055706.1 | 6 | 637 | ||
| GenBank mRNA | BC000684.2 | - | 6 | 549 | 3347335 | |
| BC007873.2 | - | 6 | 513 | 4303959 | ||
| BC071993.1 | - | 6 | 547 | |||
| BI460344.1 | - | 6 | 630 | |||
| U41745.1 | - | 6 | 524 | |||

PDAP1
Genomic Map