siRNA Details
| Gene Symbol: GAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002048.2 | NP_002039.2 | 1 | 1117 | ||
| GenBank mRNA | BC074809.2 | - | 1 | 749 | 30915530 | |
| BC074908.2 | - | 1 | 749 | 30915255 | ||
| BC132682.1 | - | 1 | 792 | 40146704 | ||
| BC136586.1 | - | 1 | 792 | 9020576 | ||
| L13698.1 | - | 1 | 1117 | |||

GAS1
Genomic Map