siRNA Details
| Gene Symbol: KRT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005556.3 | NP_005547.3 | 6 | 1092 | ||
| XM_011538325.1 | XP_011536627.1 | 6 | 1045 | |||
| XM_017019294.1 | XP_016874783.1 | 6 | 818 | |||
| GenBank mRNA | AF509887.1 | - | 6 | 1021 | ||
| AJ238246.1 | - | 6 | 1026 | |||
| AK025881.1 | - | 6 | 1019 | |||
| AK128505.1 | - | 5 | 2286 | |||
| AK128848.1 | - | 7 | 947 | |||
| AK129924.1 | - | 7 | 1026 | |||
| AK225498.1 | - | 6 | 1015 | |||
| BC002700.2 | - | 6 | 1007 | 3610347 | ||
| BC009521.1 | - | 2 | 179 | |||
| BC107082.1 | - | 3 | 351 | |||
| BC107083.1 | - | 2 | 283 | |||
| BE740515.1 | - | 2 | 223 | |||
| BE744321.1 | - | 669 | ||||
| X03212.1 | - | 6 | 1026 | |||

KRT7
Genomic Map