siRNA Details
| Gene Symbol: BST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004335.3 | NP_004326.1 | 2, 3 | 444 | ||
| GenBank mRNA | AK223124.1 | - | 2, 3 | 382 | ||
| AK291099.1 | - | 2, 3 | 389 | |||
| AK303593.1 | - | 2, 3 | 414 | |||
| BC033873.1 | - | 2, 3 | 444 | 5217945 | ||
| D28137.1 | - | 2, 3 | 372 | |||

BST2
Genomic Map