siRNA Details
| Gene Symbol: MYL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006097.4 | NP_006088.2 | 4 | 566 | ||
| NM_181526.2 | NP_852667.1 | 3 | 404 | |||
| GenBank mRNA | AF176042.1 | - | 3 | 502 | ||
| AF176043.1 | - | 2 | 340 | |||
| AK130879.1 | - | 4 | 531 | |||
| BC002648.2 | - | 3 | 343 | 3608316 | ||
| BQ953325.1 | - | 556 | ||||
| CR456843.1 | - | 3 | 462 | |||
| D82057.1 | - | 4 | 534 | |||
| J02854.1 | - | 3 | 504 | |||

MYL9
Genomic Map