siRNA Details
| Gene Symbol: ANAPC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242374.1 | NP_001229303.1 | 3 | 1293 | ||
| NM_001242375.1 | NP_001229304.1 | 3 | 878 | |||
| NM_015391.3 | NP_056206.1 | 3 | 659 | |||
| GenBank mRNA | AK001285.1 | - | 3 | 1290 | ||
| AK026550.1 | - | 3 | 613 | |||
| AL050051.1 | - | 3 | 878 | |||
| AV682398.1 | - | 3 | 624 | |||
| BC005398.1 | - | 3 | 625 | 3828623 | ||
| BX537696.1 | - | 4 | 1079 | |||

ANAPC13
Genomic Map