siRNA Details
| Gene Symbol: PRAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145201.1 | NP_001138673.1 | 2, 3 | 161 | ||
| NM_145202.4 | NP_660203.3 | 2, 3 | 161 | |||
| GenBank mRNA | AF123768.1 | - | 1, 2 | 90 | ||
| AF421885.1 | - | 2, 3 | 103 | |||
| AY158074.1 | - | 1, 2 | 91 | |||
| AY358908.1 | - | 2, 3 | 140 | |||
| BC029447.2 | - | 1, 2 | 76 | |||
| BC061643.1 | - | 2, 3 | 95 | 5748898 | ||
| BC071872.1 | - | 2, 3 | 94 | 6284088 | ||
| BC093853.1 | - | 2, 3 | 133 | 7939698 | ||
| BC101743.1 | - | 2, 3 | 133 | 8069249 | ||
| BC143590.1 | - | 2, 3 | 133 | 9052101 | ||
| BC143591.1 | - | 2, 3 | 133 | 9052102 | ||
| BC143592.1 | - | 2, 3 | 133 | 9052103 | ||
| DQ884405.1 | - | 1, 2 | 83 | |||
| GU727628.1 | - | 2, 3 | 103 | |||
| Gene Symbol: ZNF511-PRAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | EU937518.1 | - | 5, 6 | 582 | ||

PRAP1
Genomic Map