siRNA Details
| Gene Symbol: OR52E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005164.2 | NP_001005164.2 | 1 | 285 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR52E2| Gene Symbol: OR52E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005164.2 | NP_001005164.2 | 1 | 285 | ||