siRNA Details
| Gene Symbol: OR51S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004758.1 | NP_001004758.1 | 1 | 556 | ||
| GenBank mRNA | BC136932.1 | - | 1 | 632 | 9020924 | |
| BC136934.1 | - | 1 | 632 | 9020926 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR51S1| Gene Symbol: OR51S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004758.1 | NP_001004758.1 | 1 | 556 | ||
| GenBank mRNA | BC136932.1 | - | 1 | 632 | 9020924 | |
| BC136934.1 | - | 1 | 632 | 9020926 | ||