siRNA Details
| Gene Symbol: RHOD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014578.3 | NP_055393.1 | 2, 3 | 313 | ||
| GenBank mRNA | AF498973.1 | - | 2, 3 | 228 | ||
| BC001338.1 | - | 2, 3 | 281 | 3461774 | ||
| BM761353.1 | - | 2, 3 | 313 | |||
| BM761362.1 | - | 2, 3 | 313 | |||

RHOD
Genomic Map