siRNA Details
| Gene Symbol: C1orf122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142726.1 | NP_001136198.1 | 3 | 1022 | ||
| NM_198446.2 | NP_940848.2 | 3 | 1155 | |||
| GenBank mRNA | AK127381.1 | - | 3 | 659 | ||
| AY517499.1 | - | 3 | 810 | |||
| BC016402.1 | - | 2 | 2068 | |||
| BC057819.1 | - | 3 | 660 | 4792982 | ||
| BC102041.1 | - | 2 | 311 | |||
| BC107856.1 | - | 2 | 770 | |||
| BC127187.1 | - | 2 | 311 | |||
| BC127188.1 | - | 2 | 222 | |||

C1orf122
Genomic Map