siRNA Details
| Gene Symbol: PRIMA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178013.3 | NP_821092.1 | 4 | 475 | ||
| XM_011536456.2 | XP_011534758.1 | 5 | 929 | |||
| GenBank mRNA | AY225516.1 | - | 4 | 475 | ||
| AY225517.1 | - | 5 | 646 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRIMA1| Gene Symbol: PRIMA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178013.3 | NP_821092.1 | 4 | 475 | ||
| XM_011536456.2 | XP_011534758.1 | 5 | 929 | |||
| GenBank mRNA | AY225516.1 | - | 4 | 475 | ||
| AY225517.1 | - | 5 | 646 | |||