siRNA Details
| Gene Symbol: TMEM31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182541.2 | NP_872347.2 | 2 | 204 | ||
| GenBank mRNA | AK130867.1 | - | 2 | 204 | ||
| BC029575.1 | - | 2 | 205 | 5267530 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM31| Gene Symbol: TMEM31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182541.2 | NP_872347.2 | 2 | 204 | ||
| GenBank mRNA | AK130867.1 | - | 2 | 204 | ||
| BC029575.1 | - | 2 | 205 | 5267530 | ||