siRNA Details
| Gene Symbol: OR4C16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004701.2 | NP_001004701.2 | 1 | 590 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR4C16| Gene Symbol: OR4C16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004701.2 | NP_001004701.2 | 1 | 590 | ||