siRNA Details
| Gene Symbol: OR4D5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001965.1 | NP_001001965.1 | 1 | 270 | ||
| GenBank mRNA | BC136943.1 | - | 1 | 344 | 9020935 | |
| BC136945.1 | - | 1 | 344 | 9020937 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR4D5| Gene Symbol: OR4D5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001965.1 | NP_001001965.1 | 1 | 270 | ||
| GenBank mRNA | BC136943.1 | - | 1 | 344 | 9020935 | |
| BC136945.1 | - | 1 | 344 | 9020937 | ||