siRNA Details
Gene Symbol: OR10Q1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004471.2 | NP_001004471.1 | 1 | 287 | ||
GenBank mRNA | BC140731.1 | - | 1 | 287 | 9021599 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR10Q1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004471.2 | NP_001004471.1 | 1 | 287 | ||
GenBank mRNA | BC140731.1 | - | 1 | 287 | 9021599 |