siRNA Details
| Gene Symbol: OR5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004728.1 | NP_001004728.1 | 1 | 499 | ||
| GenBank mRNA | BC137060.1 | - | 1 | 524 | 9021057 | |
| BC137061.1 | - | 1 | 524 | 9021058 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR5A1| Gene Symbol: OR5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004728.1 | NP_001004728.1 | 1 | 499 | ||
| GenBank mRNA | BC137060.1 | - | 1 | 524 | 9021057 | |
| BC137061.1 | - | 1 | 524 | 9021058 | ||