siRNA Details
| Gene Symbol: OR2B6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012367.1 | NP_036499.1 | 1 | 139 | ||
| GenBank mRNA | BC109251.1 | - | 1 | 139 | 40023640 | |
| BC110457.2 | - | 1 | 139 | 40022996 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR2B6| Gene Symbol: OR2B6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012367.1 | NP_036499.1 | 1 | 139 | ||
| GenBank mRNA | BC109251.1 | - | 1 | 139 | 40023640 | |
| BC110457.2 | - | 1 | 139 | 40022996 | ||