siRNA Details
| Gene Symbol: OR1J4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004452.1 | NP_001004452.1 | 1 | 821 | ||
| GenBank mRNA | BC131606.1 | - | 1 | 820 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR1J4| Gene Symbol: OR1J4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004452.1 | NP_001004452.1 | 1 | 821 | ||
| GenBank mRNA | BC131606.1 | - | 1 | 820 | ||