siRNA Details
| Gene Symbol: OR2Z1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004699.2 | NP_001004699.1 | 1 | 656 | ||
| GenBank mRNA | BC137046.1 | - | 1 | 655 | 9021042 | |
| BC151149.1 | - | 1 | 656 | 9021043 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR2Z1| Gene Symbol: OR2Z1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004699.2 | NP_001004699.1 | 1 | 656 | ||
| GenBank mRNA | BC137046.1 | - | 1 | 655 | 9021042 | |
| BC151149.1 | - | 1 | 656 | 9021043 | ||