siRNA Details
| Gene Symbol: OR52A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012375.2 | NP_036507.2 | 1 | 466 | ||
| GenBank mRNA | BC111722.1 | - | 1 | 466 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR52A1| Gene Symbol: OR52A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012375.2 | NP_036507.2 | 1 | 466 | ||
| GenBank mRNA | BC111722.1 | - | 1 | 466 | ||