siRNA Details
| Gene Symbol: C9orf142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183241.1 | NP_899064.1 | 7 | 800 | ||
| GenBank mRNA | BC002613.2 | - | 7 | 800 | 3161564 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C9orf142| Gene Symbol: C9orf142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183241.1 | NP_899064.1 | 7 | 800 | ||
| GenBank mRNA | BC002613.2 | - | 7 | 800 | 3161564 | |