siRNA Details
| Gene Symbol: KRTAP23-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181624.1 | NP_853655.1 | 1 | 121 | ||
| GenBank mRNA | BC128435.1 | - | 1 | 125 | 40119522 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP23-1| Gene Symbol: KRTAP23-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181624.1 | NP_853655.1 | 1 | 121 | ||
| GenBank mRNA | BC128435.1 | - | 1 | 125 | 40119522 | |