siRNA Details
Gene Symbol: KRTAP19-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181608.1 | NP_853639.1 | 1 | 129 | ||
GenBank mRNA | AB180041.1 | - | 1 | 217 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KRTAP19-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181608.1 | NP_853639.1 | 1 | 129 | ||
GenBank mRNA | AB180041.1 | - | 1 | 217 |