siRNA Details
| Gene Symbol: KRTAP20-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181615.1 | NP_853646.1 | 1 | 119 | ||
| GenBank mRNA | AB096955.1 | - | 1 | 149 | ||
| BC104846.1 | - | 1 | 143 | 8143849 | ||
| BC104872.1 | - | 1 | 143 | 8143875 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP20-1| Gene Symbol: KRTAP20-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181615.1 | NP_853646.1 | 1 | 119 | ||
| GenBank mRNA | AB096955.1 | - | 1 | 149 | ||
| BC104846.1 | - | 1 | 143 | 8143849 | ||
| BC104872.1 | - | 1 | 143 | 8143875 | ||