siRNA Details
| Gene Symbol: LOC339059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120387.1 | 2 | 741 | |||
| GenBank mRNA | AK097767.1 | - | 2 | 741 | ||
| BC111769.1 | - | 1 | 788 | 40025355 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC339059| Gene Symbol: LOC339059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120387.1 | 2 | 741 | |||
| GenBank mRNA | AK097767.1 | - | 2 | 741 | ||
| BC111769.1 | - | 1 | 788 | 40025355 | ||