siRNA Details
| Gene Symbol: ADAMTSL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213604.2 | NP_998769.2 | 12 | 2425 | ||
| XM_005259549.3 | XP_005259606.1 | 13 | 2134 | |||
| XM_011527961.1 | XP_011526263.1 | 13 | 2146 | |||
| XM_011527963.1 | XP_011526265.1 | 13 | 2133 | |||
| XM_017026744.1 | XP_016882233.1 | 12 | 2268 | |||
| XM_017026745.1 | XP_016882234.1 | 12 | 2288 | |||
| GenBank mRNA | AK131571.1 | - | 12 | 2256 | ||

ADAMTSL5
Genomic Map