siRNA Details
| Gene Symbol: ANKRD34B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004441.2 | NP_001004441.2 | 5 | 688 | ||
| XM_006714599.3 | XP_006714662.1 | 5 | 450 | |||
| XM_011543366.2 | XP_011541668.1 | 5 | 492 | |||
| XM_011543369.2 | XP_011541671.1 | 3 | 448 | |||
| GenBank mRNA | BC137441.1 | - | 5 | 467 | 9021439 | |
| BC137442.1 | - | 5 | 464 | 9021440 | ||
| BC142957.1 | - | 4 | 368 | 8860359 | ||
| CR749535.1 | - | 5 | 482 | |||

ANKRD34B
Genomic Map