siRNA Details
| Gene Symbol: ZNF592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014630.2 | NP_055445.2 | 4 | 477 | ||
| XM_005254996.3 | XP_005255053.1 | 3 | 378 | |||
| XM_011522246.2 | XP_011520548.1 | 4 | 547 | |||
| XM_011522247.2 | XP_011520549.1 | 3 | 550 | |||
| XM_011522248.2 | XP_011520550.1 | 4 | 508 | |||
| XM_017022734.1 | XP_016878223.1 | 2 | 258 | |||
| GenBank mRNA | BC094688.1 | - | 4 | 429 | 30397050 | |
| BC112232.1 | - | 2 | 211 | 8327700 | ||
| BC112234.1 | - | 2 | 212 | 8327702 | ||
| BC143600.1 | - | 2 | 213 | |||
| D86966.1 | - | 4 | 711 | |||

ZNF592
Genomic Map