siRNA Details
| Gene Symbol: OR1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004450.1 | NP_001004450.1 | 1 | 72 | ||
| XM_017014695.1 | XP_016870184.1 | 2 | 601 | |||
| XM_017014696.1 | XP_016870185.1 | 2 | 633 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR1B1| Gene Symbol: OR1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004450.1 | NP_001004450.1 | 1 | 72 | ||
| XM_017014695.1 | XP_016870184.1 | 2 | 601 | |||
| XM_017014696.1 | XP_016870185.1 | 2 | 633 | |||