siRNA Details
| Gene Symbol: BMP8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181809.3 | NP_861525.2 | 7 | 1630 | ||
| GenBank mRNA | AY303954.1 | - | 7 | 1630 | ||
| BC131617.1 | - | 7 | 946 | 40116584 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BMP8A| Gene Symbol: BMP8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181809.3 | NP_861525.2 | 7 | 1630 | ||
| GenBank mRNA | AY303954.1 | - | 7 | 1630 | ||
| BC131617.1 | - | 7 | 946 | 40116584 | ||