siRNA Details
| Gene Symbol: TMEM179B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199337.2 | NP_955369.1 | 5 | 766 | ||
| XM_005273982.3 | XP_005274039.3 | 4 | 768 | |||
| GenBank mRNA | BC051355.1 | - | 5 | 741 | 6597282 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM179B| Gene Symbol: TMEM179B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199337.2 | NP_955369.1 | 5 | 766 | ||
| XM_005273982.3 | XP_005274039.3 | 4 | 768 | |||
| GenBank mRNA | BC051355.1 | - | 5 | 741 | 6597282 | |