siRNA Details
| Gene Symbol: MICAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282663.1 | NP_001269592.1 | 5 | 862 | ||
| NM_001282664.1 | NP_001269593.1 | 4 | 715 | |||
| NM_001282665.1 | NP_001269594.1 | 4 | 715 | |||
| NM_001282666.1 | NP_001269595.1 | 3 | 640 | |||
| NM_001282667.1 | NP_001269596.1 | 3 | 640 | |||
| NM_001282668.1 | NP_001269597.1 | 5 | 862 | |||
| NM_014632.3 | NP_055447.1 | 5 | 866 | |||
| XM_011520490.2 | XP_011518792.1 | 3 | 569 | |||
| XM_011520491.2 | XP_011518793.1 | 3 | 569 | |||
| XM_017018589.1 | XP_016874078.1 | 4 | 682 | |||
| XM_017018590.1 | XP_016874079.1 | 4 | 727 | |||
| XM_017018591.1 | XP_016874080.1 | 4 | 682 | |||
| GenBank mRNA | AB018293.1 | - | 5 | 827 | ||
| AB110785.1 | - | 3 | 563 | |||
| AB110786.1 | - | 3 | 563 | |||
| AK294845.1 | - | 4 | 682 | |||
| AK302893.1 | - | 5 | 944 | |||
| BC044577.1 | - | 5 | 851 | 5275364 | ||
| BX538021.1 | - | 1 | 390 | |||

MICAL2
Genomic Map