siRNA Details
| Gene Symbol: RASL11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206827.1 | NP_996563.1 | 4 | 947 | ||
| GenBank mRNA | AY439004.1 | - | 4 | 947 | ||
| BC132703.1 | - | 4 | 683 | 40146725 | ||
| BC136761.1 | - | 4 | 683 | 9020751 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RASL11A| Gene Symbol: RASL11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206827.1 | NP_996563.1 | 4 | 947 | ||
| GenBank mRNA | AY439004.1 | - | 4 | 947 | ||
| BC132703.1 | - | 4 | 683 | 40146725 | ||
| BC136761.1 | - | 4 | 683 | 9020751 | ||