siRNA Details
| Gene Symbol: OR5AN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004729.1 | NP_001004729.1 | 1 | 427 | ||
| GenBank mRNA | BC140917.1 | - | 1 | 474 | 9056729 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR5AN1| Gene Symbol: OR5AN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004729.1 | NP_001004729.1 | 1 | 427 | ||
| GenBank mRNA | BC140917.1 | - | 1 | 474 | 9056729 | |