siRNA Details
| Gene Symbol: OR7A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005190.1 | NP_001005190.1 | 1 | 902 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR7A10| Gene Symbol: OR7A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005190.1 | NP_001005190.1 | 1 | 902 | ||