siRNA Details
| Gene Symbol: PRR27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_214711.3 | NP_999876.2 | 5 | 1991 | ||
| GenBank mRNA | BX640892.1 | - | 5 | 1903 | ||
| BX640924.1 | - | 5 | 1943 | |||
| BX640997.1 | - | 5 | 1976 | |||
| BX647183.1 | - | 6 | 1812 | |||
| BX648724.1 | - | 5 | 1937 | |||
| BX648872.1 | - | 5 | 1864 | |||
| BX648895.1 | - | 5 | 1941 | |||
| BX648896.1 | - | 6 | 1876 | |||

PRR27
Genomic Map