siRNA Details
| Gene Symbol: PRR27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_214711.3 | NP_999876.2 | 3 | 408 | ||
| GenBank mRNA | BX484937.1 | - | 3 | 409 | ||
| BX640892.1 | - | 3 | 321 | |||
| BX640924.1 | - | 3 | 359 | |||
| BX640997.1 | - | 3 | 395 | |||
| BX647183.1 | - | 3 | 280 | |||
| BX647185.1 | - | 4 | 398 | |||
| BX648724.1 | - | 3 | 357 | |||
| BX648872.1 | - | 3 | 280 | |||
| BX648895.1 | - | 3 | 357 | |||
| BX648896.1 | - | 4 | 292 | |||

PRR27
Genomic Map