siRNA Details
| Gene Symbol: KLHL31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003760.4 | NP_001003760.2 | 2 | 720 | ||
| XM_005249107.4 | XP_005249164.1 | 2 | 915 | |||
| XM_017010858.1 | XP_016866347.1 | 2 | 1830 | |||
| GenBank mRNA | BC137267.1 | - | 1 | 605 | 9021264 | |
| BC137269.1 | - | 1 | 605 | 9021266 | ||
| EF633513.1 | - | 2 | 720 | |||

KLHL31
Genomic Map